Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Benzyloxycarbonyl-L-ornithine, 98%

Catalog No. AAH6305206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6305206 5 g
AAH6305203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6305206 Supplier Thermo Scientific Chemicals Supplier No. H6305206

missing translation for 'validMainframeMsg'

CAS: 2640-58-6 | C13H18N2O4 | 266.297 g/mol

Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.

Solubility
Soluble in 0.1N NaOG (1 mg/ml-clear, colorless solution). Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2640-58-6
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00038200
InChI Key ZYGRWJVRLNJIMR-NSHDSACASA-N
Synonym z-orn-oh, cbz-l-ornithine, cbz-orn-oh, l-ornithine, n2-phenylmethoxy carbonyl, 2s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, s-5-amino-2-benzyloxycarbonylamino pentanoic acid, s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, pubchem18987, carbobenzyloxy-l-ornithine, na-carbobenzoxy-l-ornithine
PubChem CID 7010490
IUPAC Name (2S)-5-amino-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCN)C(=O)O

Specifications

Color White
Solubility Information Soluble in 0.1N NaOG (1mg/ml-clear,colorless solution). Slightly soluble in water.
Quantity 5 g
Formula Weight 266.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-ornithine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.