Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-serine, 98%, Thermo Scientific™

Catalog No. AAH6372806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6372806 5 g
AAH6372803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6372806 Supplier Thermo Scientific Chemicals Supplier No. H6372806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65806-90-8
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00077038
InChI Key TXDGEONUWGOCJG-GFCCVEGCSA-N
Synonym z-d-ser tbu-oh, cbz-o-tert-butyl-d-serine, r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-cbz-o-tert-butyl-d-serine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, ambotzzaa1152, n-alpha-benzyloxycarbonyl-o-t-butyl-d-serine, n-alpha-carbobenzoxy-d-serine t-butyl ether, r-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-d-serine
PubChem CID 1715619
IUPAC Name (2R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 295.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-serine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.