Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Boc-D-asparagine, 95%

Catalog No. AAH6112106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6112106 5 g
AAH6112114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6112106 Supplier Thermo Scientific Chemicals Supplier No. H6112106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 75647-01-7 | C9H16N2O5 | 232.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75647-01-7
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00065558
InChI Key FYYSQDHBALBGHX-RXMQYKEDSA-N
Synonym boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid
PubChem CID 1623168
IUPAC Name (2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

Specifications

Color White
Beilstein 4843040
Quantity 5 g
Formula Weight 232.23
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-D-asparagine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.