Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Boc-L-ornithine, 95%

Catalog No. AAH6201414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6201414 25 g
AAH6201406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6201414 Supplier Thermo Scientific Chemicals Supplier No. H6201414
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 21887-64-9 | C10H20N2O4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21887-64-9
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD00076970
InChI Key AMPVNPYPOOQUJF-ZETCQYMHSA-N
Synonym boc-orn-oh, boc-l-ornithine, n, a-boc-l-ornithine, l-ornithine, n2-1,1-dimethylethoxy carbonyl, 2s-5-amino-2-tert-butoxycarbonyl amino pentanoic acid, n2-boc-l-ornithine, nalpha-boc-l-ornithine, pubchem18983, nalpha-boc-l-ornithine;
PubChem CID 7018784
IUPAC Name (2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCN)C(=O)O

Specifications

Melting Point 207°C to 208°C
Color White
Beilstein 4313353
Quantity 25 g
Formula Weight 232.28
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-L-ornithine
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.