Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Boc-Nepsilon-(2-chlorobenzyloxycarbonyl)-D-lysine, 98%, Thermo Scientific™

Catalog No. AAH6306906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6306906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6306906 Supplier Thermo Scientific Chemicals Supplier No. H6306906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57096-11-4
Molecular Formula C19H27ClN2O6
Molecular Weight (g/mol) 414.88
MDL Number MFCD00065585
InChI Key ATUMDPHEFWGCJF-GGYSOQFKNA-N
Synonym boc-d-lys 2-cl-z-oh, boc-n'-2-chloro-cbz-d-lysine, boc-d-lys 2-cl-cbz-oh, ambotzbaa1036, boc-d-lys 2-chloro-z-oh, boc-l-lys 2-cl-cbz-oh, n, a-boc-n, a-2-chloro-z-d-lysine, boc-d-lys 2-cl-z-oh tlc, n-boc-n'-2-chlorobenzyloxy carbonyl-d-lysine, n-tert-butoxy carbonyl-n'-2-chlorobenzyloxycarbonyl-d-lysine
PubChem CID 7269325
SMILES CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1Cl)C(O)=O

Specifications

Color White
Beilstein 5913148
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-6-({[(2-chlorophenyl)methoxy]carbonyl}amino)hexanoic acid
Quantity 5 g
Formula Weight 414.88
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-Nepsilon-(2-chlorobenzyloxycarbonyl)-D-lysine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.