Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Boc-Nepsilon-formyl-L-lysine, 95%, Thermo Scientific™

Catalog No. AAH6663506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6663506 5 g
AAH6663503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6663506 Supplier Thermo Scientific Chemicals Supplier No. H6663506

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2483-47-8
Molecular Formula C12H22N2O5
Molecular Weight (g/mol) 274.32
MDL Number MFCD00037361
InChI Key QYCPNUMTVZBTMM-UHFFFAOYNA-N
Synonym boc-lys for-oh, s-2-tert-butoxycarbonyl amino-6-formamidohexanoic acid, l-lysine, n2-1,1-dimethylethoxy carbonyl-n6-formyl, 2s-2-tert-butoxycarbonyl amino-6-formamidohexanoic acid, boc-l-lys epsilon-formyl-oh, n, a-boc-n, a-formyl-l-lysine, 2s-6-formamido-2-2-methylpropan-2-yl oxycarbonylamino hexanoic acid
PubChem CID 1581890
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-6-formamidohexanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCCNC=O)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 274.32
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-Nepsilon-formyl-L-lysine
EINECSNumber 219-619-4
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.