Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine, 95%, Thermo Scientific Chemicals

Catalog No. AAH62997MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62997MD 250 mg
AAH6299703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62997MD Supplier Thermo Scientific Chemicals Supplier No. H62997MD

missing translation for 'validMainframeMsg'

CAS: 186698-61-3 | Molecular Formula: C24H38N4O7S | Molecular Weight: 526.649 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186698-61-3
Molecular Formula C24H38N4O7S
Molecular Weight (g/mol) 526.649
MDL Number MFCD00236822
InChI Key CVFXPOKENLGCID-QGZVFWFLSA-N
Synonym boc-d-arg pbf-oh, boc-nw-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine, ambotzbaa1021, 2r-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2r-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid, e-n∼5∼-amino 2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonyl amino methylidene-n∼2∼-tert-butoxycarbonyl-d-ornithine, n-alpha-t-butyloxycarbonyl-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine
PubChem CID 16213160
IUPAC Name (2R)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C

Specifications

Quantity 250 mg
Formula Weight 526.65
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.