Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Carbobenzoxy-D-asparagine 99.0+%, TCI America™
SDP

Catalog No. C0666100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0666100MG 100 mg
C06661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0666100MG Supplier TCI America Supplier No. C0666100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4474-86-6
Molecular Formula C12H14N2O5
Molecular Weight (g/mol) 266.25
MDL Number MFCD00065696
InChI Key FUCKRCGERFLLHP-UHFFFAOYNA-N
Synonym n-cbz-d-asparagine, z-d-asn-oh, nalpha-cbz-d-asparagine, z-d-asparagine, cbz d-asparagine, cbz-d-asn-oh, n-carbobenzoxy-d-asparagine, n-cbz-d-asn-oh, nalpha-carbobenzoxy-d-asparagine, 2r-2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid
PubChem CID 1712147
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoic acid
SMILES NC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 165°C
Color White
Quantity 100 mg
Formula Weight 266.25
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-D-asparagine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.