Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Carbobenzoxy-D-histidine 98.0+%, TCI America™
SDP

Catalog No. C21331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21331G 1 g
C21335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21331G Supplier TCI America Supplier No. C21331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67424-93-5
Molecular Formula C14H15N3O4
Molecular Weight (g/mol) 289.291
MDL Number MFCD00067702
InChI Key WCOJOHPAKJFUDF-GFCCVEGCSA-N
Synonym z-d-his-oh, nalpha-cbz-d-histidine, cbz-d-his-oh, cbz-d-histidine, z-d-histidine, nalpha-carbobenzoxy-d-histidine, n alpha-cbz-d-histidine, n alpha-benzyloxycarbonyl-d-histidine, n-benzyloxy carbonyl-d-histidine, d-histidine,n-phenylmethoxy carbonyl
PubChem CID 736161
IUPAC Name (2R)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O

Specifications

Melting Point 163°C
Quantity 1 g
Formula Weight 289.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-D-histidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.