Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Carbobenzoxy-L-arginine 97.0+%, TCI America™
SDP

Catalog No. C07571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07571G 1 g
C075725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07571G Supplier TCI America Supplier No. C07571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1234-35-1
Molecular Formula C14H20N4O4
Molecular Weight (g/mol) 308.338
MDL Number MFCD00001762
InChI Key SJSSFUMSAFMFNM-NSHDSACASA-N
Synonym z-arg-oh, nalpha-cbz-l-arginine, nalpha-carbobenzyloxy-l-arginine, cbz-arg-oh, z-l-arg-oh, cbz-l-arginine, cbz-l-arg-oh, n-benzyloxycarbonyl-l-arginine, carbobenzoxy-l-arginine, z-l-arginine
PubChem CID 71055
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N)C(=O)O

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 308.34
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-L-arginine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.