Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Fmoc-N^d-trityl-D-glutamine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6253906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6253906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6253906 Supplier Thermo Scientific Chemicals Supplier No. H6253906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 200623-62-7 | Molecular Formula: C39H34N2O5 | Molecular Weight: 610.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200623-62-7
Molecular Formula C39H34N2O5
Molecular Weight (g/mol) 610.71
MDL Number MFCD00151924
InChI Key WDGICUODAOGOMO-PGUFJCEWSA-N
Synonym fmoc-d-gln trt-oh, n-fmoc-n'-trityl-d-glutamine, fmoc-gln trt, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, ambotzfaa1322, pubchem12399, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-trityl-d-glutamine, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n-triphenylmethyl-d-glutamine, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-triphenylmethyl carbamoyl butanoic acid
PubChem CID 24820181
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Quantity 5 g
Formula Weight 610.7
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Ndelta-trityl-D-glutamine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.