Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Glycyl-D-asparagine 98.0+%, TCI America™
SDP

Catalog No. G01161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01161G Supplier TCI America Supplier No. G01161G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24667-21-8
Molecular Formula C6H11N3O4
Molecular Weight (g/mol) 189.171
MDL Number MFCD00069939
InChI Key FUESBOMYALLFNI-GSVOUGTGSA-N
Synonym H-Gly-D-Asn-OH
PubChem CID 1552085
IUPAC Name (2R)-4-amino-2-[(2-aminoacetyl)amino]-4-oxobutanoic acid
SMILES C(C(C(=O)O)NC(=O)CN)C(=O)N

Specifications

Color White
Quantity 1 g
Formula Weight 189.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Glycyl-D-asparagine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.