Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Glycyl-DL-asparagine 98.0+%, TCI America™
SDP

Catalog No. G01171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01171G Supplier TCI America Supplier No. G01171G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crystalline powder with High- purity for laboratory applications

Chemical Identifiers

CAS 32729-21-8
Molecular Formula C6H11N3O4
Molecular Weight (g/mol) 189.171
MDL Number MFCD00038155
InChI Key FUESBOMYALLFNI-UHFFFAOYSA-N
Synonym H-Gly-DL-Asn-OH
PubChem CID 306141
IUPAC Name 4-amino-2-[(2-aminoacetyl)amino]-4-oxobutanoic acid
SMILES C(C(C(=O)O)NC(=O)CN)C(=O)N

Specifications

Color White
Quantity 1 g
Formula Weight 189.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Glycyl-DL-asparagine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.