Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nalpha,Nalpha-Bis(Carboxymethyl)-L-Lysine Hydrate

Catalog No. 145805G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-580-5G 5 g
14-580-1G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-580-5G Supplier Sigma Aldrich Supplier No. 145805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tricarboxylic acid used for preparing chelating resins employed for the chromatography of proteins

Specifications

CAS 941689-36-7
Molecular Formula C10H18N2O6 -+ xH2O
Linear Formula C10H18N2O6 -+ xH2O
MDL Number MFCD24387945
Quantity 5 g
Synonym (S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid
Molecular Weight (g/mol) 262.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.