Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%

Catalog No. AAH6278514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6278514 25 g
AAH6278522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6278514 Supplier Thermo Scientific Chemicals Supplier No. H6278514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 15098-69-8 | C28H53N3O6 | 527.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15098-69-8
Molecular Formula C28H53N3O6
Molecular Weight (g/mol) 527.75
MDL Number MFCD00038892
InChI Key HRLHJTYAMCGERD-VAMKTSTMNA-N
Synonym boc-lys boc-oh.dcha, boc-lys boc-oh dcha, dicyclohexylamine s-2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine dicyclohexylamine salt, boc-lys boc-oh dicyclohexammonium salt, n,n'-bis tert-butoxycarbonyl-l-lysine dicyclohexylamine 1:1, 2s-2,6-bis tert-butoxycarbonyl amino hexanoic acid; dicha, boc-lys boc-ohcha, boc-lys boc-oh?cha, boc-lys boc-ohdcha
PubChem CID 12017189
IUPAC Name (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid;N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 139°C to 143°C
Color White
Beilstein 6262378
Quantity 25 g
Formula Weight 527.74
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt
EINECSNumber 239-150-9
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.