Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-L-glutamine 98.0+%, TCI America™
SDP

Catalog No. B164925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B164925G 25 g
B164910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B164925G Supplier TCI America Supplier No. B164925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13726-85-7
Molecular Formula C10H17N2O5
Molecular Weight (g/mol) 245.26
MDL Number MFCD00065571
InChI Key VVNYDCGZZSTUBC-LURJTMIESA-M
Synonym boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamine
PubChem CID 83687
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-carbamoylbutanoate
SMILES CC(C)(C)OC(=O)N[C@@H](CCC(N)=O)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 246.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-L-glutamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.