Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-L-tryptophan N-Succinimidyl Ester, TCI America™
SDP

Catalog No. B34355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34355G Supplier TCI America Supplier No. B34355G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3392-11-8
Molecular Formula C20H23N3O6
Molecular Weight (g/mol) 401.42
MDL Number MFCD00022756
InChI Key CPJXMXQYRHNIFU-UHFFFAOYNA-N
Synonym Nalpha-Boc-L-tryptophan N-Succinimidyl Ester, Boc-Trp-OSu
PubChem CID 98767
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-3-(1H-indol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)ON3C(=O)CCC3=O

Specifications

Melting Point 145°C
Color White
Quantity 5 g
Formula Weight 401.42
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-L-tryptophan N-Succinimidyl Ester
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.