Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-N1-formyl-L-tryptophan 98.0+%, TCI America™
SDP

Catalog No. B22605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22605G Supplier TCI America Supplier No. B22605G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 47355-10-2
Molecular Formula C17H20N2O5
Molecular Weight (g/mol) 332.36
MDL Number MFCD00065992
InChI Key IHXHBYFWSOYYTR-ZDUSSCGKSA-N
Synonym boc-trp for-oh, n-tert-butoxycarbonyl-n'-formyl-l-tryptophan, chembl63415, nalpha-tert-butoxycarbonyl-n1-formyl-l-tryptophan, s-2-tert-butoxycarbonyl amino-3-1-formyl-1h-indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-formylindol-3-yl propanoic acid, boc-trp cho-oh, pubchem12191, t-boc-trp cho-oh, nalpha-boc-n1-formyl-l-tryptophan
PubChem CID 7017963
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1-formyl-1H-indol-3-yl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CN(C=O)C2=CC=CC=C12)C(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 332.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-N1-formyl-L-tryptophan
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.