Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-Nomega-(p-toluenesulfonyl)-L-arginine 95.0+%, TCI America™
SDP

Catalog No. B37665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37665G Supplier TCI America Supplier No. B37665G

Chemical Identifiers

CAS 13836-37-8
Molecular Formula C18H28N4O6S
Molecular Weight (g/mol) 428.504
MDL Number MFCD00065557
InChI Key WBIIPXYJAMICNU-AWEZNQCLSA-N
Synonym boc-arg tos-oh, boc-arg tos, n alpha-boc-n omega-tosyl-l-arginine, n 2-tert-butoxycarbonyl-n g-tosyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-4-methylbenzenesulfonyl carbamimidamido pentanoic acid, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5-imino 4-methylphenyl sulfonyl amino methyl, boc-arg tos oh, boc-arg ts, ambotzbaa1068, boc arg tos-oh
PubChem CID 83767
IUPAC Name (2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N

Specifications

Melting Point 105°C
Color White
Quantity 5 g
Formula Weight 428.50
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-Nomega-(p-toluenesulfonyl)-L-arginine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.