Learn More
Description
Naltrexone Hydrochloride USP is a white to almost white powder. It is very hygroscopic. Naltrexone Hydrochloride is very soluble in water and slightly soluble in alcohol. Water content may be up to 11% for the dihydrate version.
Alternative name(s): Naltrexone hydrochloride dihydrate
Specifications
Specifications
| Chemical Name or Material | Naltrexone Hydrochloride, USP (Dihydrate) |
| Packaging | Jar 30ml, White, HDPE, Round, 28-400, TRIVENI-DB, CT |
| CAS | 850808-02-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 413.8952 |
| Quantity | 5 g |
| Percent Purity | 98 to 102 % (on anhydrous, solvent free basis) |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
