Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Naltrexone Hydrochloride, USP (Dihydrate)
SDP

Catalog No. 14186156 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-156 100 g
14-185-962 5 g
14-186-056 1 g
14-185-485 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-186-156 Supplier Medisca Supplier No. 088705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naltrexone Hydrochloride USP is a white crystalline powder. It is very hygroscopic.

Naltrexone Hydrochloride USP is a white to almost white powder. It is very hygroscopic. Naltrexone Hydrochloride is very soluble in water and slightly soluble in alcohol. Water content may be up to 11% for the dihydrate version.

Alternative name(s): Naltrexone hydrochloride dihydrate

Specifications

Chemical Name or Material Naltrexone Hydrochloride, USP (Dihydrate)
Packaging Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT
CAS 850808-02-5
Type Drug Standard
Molecular Weight (g/mol) 413.8952
Quantity 100 g
Percent Purity 98 to 102 % (on anhydrous, solvent free basis)
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.