Learn More
Medchemexpress LLC Naltriben mesylate | 122517-78-6 | 99.7% | 511.59 g/mol | C27H29NO7S | 5 MG
SDP
Catalog No. 5000213457
Encompass_Preferred
Naltriben mesylate is the methanesulfonate salt of naltriben, a selective δ2-opioid receptor antagonist and TRPM7 activator used in biochemical and pharmacological research. The compound is supplied as a white to off-white solid and is accompanied by a datasheet and certificate of analysis for identity and quality assurance.
  • Selective δ2-opioid receptor antagonist and TRPM7 activator.
  • High purity; LCMS purity 99.7% per COA.
  • Provided as a mesylate salt for improved stability and handling.
  • Characterized molecular formula C27H29NO7S and molecular weight 511.59 g/mol.
  • Available in small research quantities suitable for laboratory assays.
  • White to off-white solid with analytical data consistent with structure.
Catalog No. 50-002-13457 Supplier Medchemexpress LLC Supplier No. HY1013025MG
Naltriben mesylate is the methanesulfonate salt of naltriben, a selective δ2-opioid receptor antagonist and TRPM7 activator used in biochemical and pharmacological research. The compound is supplied as a white to off-white solid and is accompanied by a datasheet and certificate of analysis for identity and quality assurance.
  • Selective δ2-opioid receptor antagonist and TRPM7 activator.
  • High purity; LCMS purity 99.7% per COA.
  • Provided as a mesylate salt for improved stability and handling.
  • Characterized molecular formula C27H29NO7S and molecular weight 511.59 g/mol.
  • Available in small research quantities suitable for laboratory assays.
  • White to off-white solid with analytical data consistent with structure.