Learn More
Cayman Chemical NaltrIndolehydrochlorIde 10mg
SDP
Catalog No. 502894403
Encompass_Preferred
A potent antagonist of the human δ-opioid receptor (Ki = 0.02-0.3 nM), with much lower affinities for κ- and µ-opioid receptors (Kis = 10-66 and 6-64 nM, respectively)
Catalog No. 50-289-4403 Supplier Cayman Chemical Supplier No. 900070510 MG
A potent antagonist of the human δ-opioid receptor (Ki = 0.02-0.3 nM), with much lower affinities for κ- and µ-opioid receptors (Kis = 10-66 and 6-64 nM, respectively)