Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naluzotan hydrochloride | 740873-82-9 | 99.6% | 487.10 g/mol | C23H39ClN4O3S | 10 MG
SDP

Catalog No. 5000286679
Encompass_Preferred

Naluzotan hydrochloride is the hydrochloride salt of a potent, selective amidosulfonamide 5-HT1A receptor agonist used in preclinical research. It is supplied as a solid research chemical with characterized purity and analytical data, intended for laboratory use only.

  • High purity (99.6%) suitable for research applications.
  • Potent, selective 5-HT1A receptor agonist (IC50 ≈ 20 nM; Ki ≈ 5.1 nM).
  • Weak hERG K+ channel blocker (IC50 ≈ 3,800 nM), relevant for cardiac safety studies.
  • Solid form; molecular weight 487.10 g/mol; formula C23H39ClN4O3S.
  • Supplied in a 10 mg research pack and for research use only.

Catalog No. 50-002-86679 Supplier Medchemexpress LLC Supplier No. HY14848A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naluzotan hydrochloride is the hydrochloride salt of a potent, selective amidosulfonamide 5-HT1A receptor agonist used in preclinical research. It is supplied as a solid research chemical with characterized purity and analytical data, intended for laboratory use only.

  • High purity (99.6%) suitable for research applications.
  • Potent, selective 5-HT1A receptor agonist (IC50 ≈ 20 nM; Ki ≈ 5.1 nM).
  • Weak hERG K+ channel blocker (IC50 ≈ 3,800 nM), relevant for cardiac safety studies.
  • Solid form; molecular weight 487.10 g/mol; formula C23H39ClN4O3S.
  • Supplied in a 10 mg research pack and for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.