Learn More
Cayman Chemical NAmIdInoLaspartIc AcId 100mg
SDP
Catalog No. 502950818
Encompass_Preferred
A guanidino compound with epileptogenic activity; induces whole-cell currents in rat hippocampal slices; intrahippocampal administration induces status epilepticus and neuronal cell death in rats
Catalog No. 50-295-0818 Supplier Cayman Chemical Supplier No. 34092100 MG
A guanidino compound with epileptogenic activity; induces whole-cell currents in rat hippocampal slices; intrahippocampal administration induces status epilepticus and neuronal cell death in rats