Learn More
Cayman Chemical N-Amidino-L-aspartic Acid, Purity: ≥95%, CAS: 6133-30-8, 500 mg
SDP
Catalog No. 502946518
Encompass_Preferred
A guanidino compound with epileptogenic activity; induces whole-cell currents in rat hippocampal slices; intrahippocampal administration induces status epilepticus and neuronal cell death in rats
Catalog No. 50-294-6518 Supplier Cayman Chemical Supplier No. 34092500 MG
A guanidino compound with epileptogenic activity; induces whole-cell currents in rat hippocampal slices; intrahippocampal administration induces status epilepticus and neuronal cell death in rats