Learn More
Cayman Chemical Neamine (hydrochloride), Purity: ≥95%, CAS: 15446-43-2, 50 mg
SDP
Catalog No. 502986282
Encompass_Preferred
A natural degradation product of neomycin; a common core molecule used in the synthesis of aminoglycoside antibiotics
Catalog No. 50-298-6282 Supplier Cayman Chemical Supplier No. 2175250 MG
A natural degradation product of neomycin; a common core molecule used in the synthesis of aminoglycoside antibiotics