Learn More
Medchemexpress LLC Namitecan (ST-1968) | 372105-27-6 | 98.0% | 434.44 | 50 MG
SDP
Catalog No. 5000362927
Encompass_Preferred
Namitecan is a potent topoisomerase I inhibitor with antitumor properties. It is intended for research use only and not for sale to patients.
  • Exhibits antitumor activity.
  • Acts as a potent topoisomerase I inhibitor.
  • Inhibits EGFR expression in various cell lines.
  • Induces apoptosis in A431 and A431/TPT cells.
  • Shows cytotoxicity against NCI-H460 and H460 NSCLC cells.
  • Demonstrates synergistic antitumor effects in SCC models when combined with cetuximab.
  • Achieves complete response rate in mice with A431 tumors.
  • Soluble in DMSO (250 mg/mL).
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years.
  • Can be stored in solvent at -80°C for 2 years or -20°C for 1 year.
Catalog No. 50-003-62927 Supplier Medchemexpress LLC Supplier No. HY1482150MG
Namitecan is a potent topoisomerase I inhibitor with antitumor properties. It is intended for research use only and not for sale to patients.
  • Exhibits antitumor activity.
  • Acts as a potent topoisomerase I inhibitor.
  • Inhibits EGFR expression in various cell lines.
  • Induces apoptosis in A431 and A431/TPT cells.
  • Shows cytotoxicity against NCI-H460 and H460 NSCLC cells.
  • Demonstrates synergistic antitumor effects in SCC models when combined with cetuximab.
  • Achieves complete response rate in mice with A431 tumors.
  • Soluble in DMSO (250 mg/mL).
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years.
  • Can be stored in solvent at -80°C for 2 years or -20°C for 1 year.