Learn More
AdipoGen Nampt (Visfatin/PBEF) (human) (IntraCellular) ELISA Kit (96 wells)
SDP
Supplier:  AdipoGen AG45A0008YEKKI01
Encompass_Preferred
ELISA Assay. Detects human Nampt. Weakly cross-reacts (<15%) with mouse Nampt and rat Nampt. Does not cross-react with human adiponectin, human resistin, human RELM-beta or human leptin. Colorimetric assay. Sample Types: Cell Lysate. Range: 0.25 to 16ng/ml. Sensitivity: 30pg/ml. Nicotinamide phosphoribosyltransferase (Nampt; PBEF; Visfatin) is involved in the biosynthesis of nicotinamide adenine dinucleotide (NAD+). Two forms of Nampt exist, an intracellular form (iNampt) and an extracellular form (eNampt). While the function of iNampt as an essential and rate-limiting NAD+ biosynthetic enzyme is well established, the physiological role of eNampt is still a matter of debate. It has been shown that Nampt expression is increased in different diseases, like diabetes, obesity, fetal growth retardation, sepsis, inflammatory bowel disease, Crohn’s disease and rheumatoid arthritis. Furthermore, Nampt might be a marker of pre-eclampsia and of endothelial health.
Catalog No. 50-205-3308