Learn More
Medchemexpress LLC Nampt-IN-10 (TFA) | 2567724-20-1 | >98.6% | 701.59 | 25 MG
SDP
Catalog No. 5000362583
Encompass_Preferred
Nampt-IN-10 TFA (compound 4) is a Nicotinamide Phosphoribosyltransferase (NAMPT) inhibitor. It shows cellular potency to A2780 and CORL23 cell lines with IC50s of 5 and 19 nM, respectively. Nampt-IN-10 TFA can be used as a novel non-antimitotic payload for ADCs.
  • Shows cellular potency to A2780, CORL23, and c-Kit and HER2 expressing cell lines.
  • IC50 values are 5 nM (A2780), 19 nM (CORL23), 2 nM (NCI-H526 with c-Kit expressing), 0.4 nM (MDA-MB453 with HER2 expressing), and 1 nM (NCI-N87 with HER2 expressing).
  • Can be used as a novel non-antimitotic payload for ADCs.
Catalog No. 50-003-62583 Supplier Medchemexpress LLC Supplier No. 25MGHY147193
Nampt-IN-10 TFA (compound 4) is a Nicotinamide Phosphoribosyltransferase (NAMPT) inhibitor. It shows cellular potency to A2780 and CORL23 cell lines with IC50s of 5 and 19 nM, respectively. Nampt-IN-10 TFA can be used as a novel non-antimitotic payload for ADCs.
  • Shows cellular potency to A2780, CORL23, and c-Kit and HER2 expressing cell lines.
  • IC50 values are 5 nM (A2780), 19 nM (CORL23), 2 nM (NCI-H526 with c-Kit expressing), 0.4 nM (MDA-MB453 with HER2 expressing), and 1 nM (NCI-N87 with HER2 expressing).
  • Can be used as a novel non-antimitotic payload for ADCs.