Learn More
Medchemexpress LLC Nampt-IN-10 (TFA) | 2567724-20-1 | 98.6% | C27H28FN5O2·C2HF3O2 | 10MG
SDP
Catalog No. 5000258821
Encompass_Preferred
Nampt-IN-10 TFA is the trifluoroacetate salt of a potent nicotinamide phosphoribosyltransferase (NAMPT) inhibitor provided for research use; it exhibits cellular activity and is supplied at high purity for in vitro and preclinical studies.
  • Trifluoroacetate salt form for improved handling and solubility
  • High purity (98.6%) suitable for sensitive assays
  • Demonstrated cellular potency in cancer cell lines
  • Supplied in multiple pack sizes to suit experimental needs
  • For research use only; not for clinical or diagnostic use
Catalog No. 50-002-58821 Supplier Medchemexpress LLC Supplier No. HY14719310MG
Nampt-IN-10 TFA is the trifluoroacetate salt of a potent nicotinamide phosphoribosyltransferase (NAMPT) inhibitor provided for research use; it exhibits cellular activity and is supplied at high purity for in vitro and preclinical studies.
  • Trifluoroacetate salt form for improved handling and solubility
  • High purity (98.6%) suitable for sensitive assays
  • Demonstrated cellular potency in cancer cell lines
  • Supplied in multiple pack sizes to suit experimental needs
  • For research use only; not for clinical or diagnostic use