Learn More
Medchemexpress LLC Nampt-IN-3 | 2121591-52-2 | 99.0% | 503.55 | 10 MG
SDP
Catalog No. 5000344796
Encompass_Preferred
Nampt-IN-3 (Compound 35) is an agent that simultaneously inhibits nicotinamide phosphoribosyltransferase (NAMPT) and Histone Deacetylase (HDAC) with IC50 values of 31 nM and 55 nM, respectively. It effectively induces cell apoptosis and autophagy, ultimately leading to cell death. This product is intended for research use only and is not sold to patients. It appears as a white to off-white solid.
  • Simultaneously inhibits NAMPT and HDAC.
  • Induces cell apoptosis and autophagy.
  • Leads to cell death.
  • High purity of 98.97%.
  • Available in both solid and pre-dissolved solution forms for convenience.
  • Used in research for cytotoxicity studies in various cell lines such as HCT-116, HepG2, and MDA-MB-231.
Catalog No. 50-003-44796 Supplier Medchemexpress LLC Supplier No. HY108701100MG
Nampt-IN-3 (Compound 35) is an agent that simultaneously inhibits nicotinamide phosphoribosyltransferase (NAMPT) and Histone Deacetylase (HDAC) with IC50 values of 31 nM and 55 nM, respectively. It effectively induces cell apoptosis and autophagy, ultimately leading to cell death. This product is intended for research use only and is not sold to patients. It appears as a white to off-white solid.
  • Simultaneously inhibits NAMPT and HDAC.
  • Induces cell apoptosis and autophagy.
  • Leads to cell death.
  • High purity of 98.97%.
  • Available in both solid and pre-dissolved solution forms for convenience.
  • Used in research for cytotoxicity studies in various cell lines such as HCT-116, HepG2, and MDA-MB-231.