Learn More
Medchemexpress LLC Nampt-IN-3 | 2121591-52-2 | MFCD28053555 | 99.0% | 503.55 g/mol | C29H25N7O2 | 5 MG
SDP
Catalog No. 5000263382
Encompass_Preferred
Nampt-IN-3 is a high-purity, research-grade small molecule inhibitor that targets nicotinamide phosphoribosyltransferase (NAMPT) and histone deacetylases (HDAC). Supplied as a white to off-white solid, it exhibits low-nanomolar biochemical potency and documented cellular activity suitable for preclinical studies.
  • High purity (98.97%) solid suitable for research use.
  • Dual inhibitory activity against NAMPT and HDAC (IC50s ~31 nM and 55 nM).
  • Documented cytotoxicity in multiple cell lines (example IC50s in μM range).
  • Soluble in DMSO at high concentration; ultrasonic assistance recommended.
  • Stable as powder and in solution when stored per recommendations.
  • Available in small research pack sizes for early-stage studies.
Catalog No. 50-002-63382 Supplier Medchemexpress LLC Supplier No. HY1087015MG
Nampt-IN-3 is a high-purity, research-grade small molecule inhibitor that targets nicotinamide phosphoribosyltransferase (NAMPT) and histone deacetylases (HDAC). Supplied as a white to off-white solid, it exhibits low-nanomolar biochemical potency and documented cellular activity suitable for preclinical studies.
  • High purity (98.97%) solid suitable for research use.
  • Dual inhibitory activity against NAMPT and HDAC (IC50s ~31 nM and 55 nM).
  • Documented cytotoxicity in multiple cell lines (example IC50s in μM range).
  • Soluble in DMSO at high concentration; ultrasonic assistance recommended.
  • Stable as powder and in solution when stored per recommendations.
  • Available in small research pack sizes for early-stage studies.