Learn More
Medchemexpress LLC Nampt-IN-5 | 2380013-17-0 | 98.0% | 100 MG
SDP
Catalog No. 5000341122
Encompass_Preferred
Nampt-IN-5 is a potent inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) and also inhibits CYP3A4 activity. It demonstrates significant antiproliferative activity, with cellular IC50s of 0.7 nM against A2780 cells and 3.9 nM against COR-L23 cells.
  • Potent NAMPT inhibitor
  • Inhibits CYP3A4 activity
  • Exhibits cellular IC50 of 0.7 nM against A2780 cells
  • Exhibits cellular IC50 of 3.9 nM against COR-L23 cells
  • Soluble in DMSO at 50 mg/mL
  • Stable as powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-003-41122 Supplier Medchemexpress LLC Supplier No. HY130606100MG
Nampt-IN-5 is a potent inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) and also inhibits CYP3A4 activity. It demonstrates significant antiproliferative activity, with cellular IC50s of 0.7 nM against A2780 cells and 3.9 nM against COR-L23 cells.
  • Potent NAMPT inhibitor
  • Inhibits CYP3A4 activity
  • Exhibits cellular IC50 of 0.7 nM against A2780 cells
  • Exhibits cellular IC50 of 3.9 nM against COR-L23 cells
  • Soluble in DMSO at 50 mg/mL
  • Stable as powder for 3 years at -20°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C