Learn More
Medchemexpress LLC Nampt-IN-5 | 2380013-17-0 | 98.0% | C25H25N5O2 | 10MG
SDP
Catalog No. 5000259972
Encompass_Preferred
Nampt-IN-5 is a research-stage small molecule inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) that demonstrates potent antiproliferative activity in cancer cell lines and has documented metabolic interaction with CYP3A4. The compound is characterized by a defined purity and recommended solvent/formulation guidance for both in vitro and in vivo studies, and is intended for research use only.
  • Potent nicotinamide phosphoribosyltransferase (NAMPT) inhibitor with sub-nanomolar cellular activity.
  • Demonstrated cellular IC50s of 0.7 nM (A2780) and 3.9 nM (COR-L23).
  • Inhibits CYP3A4, enabling metabolic interaction studies.
  • High purity (98.02%) suitable for research applications.
  • Soluble in DMSO and compatible with recommended in vivo formulations.
  • Stable as a powder at -20°C; follow solvent storage guidelines for solutions.
Catalog No. 50-002-59972 Supplier Medchemexpress LLC Supplier No. HY13060610MG
Nampt-IN-5 is a research-stage small molecule inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) that demonstrates potent antiproliferative activity in cancer cell lines and has documented metabolic interaction with CYP3A4. The compound is characterized by a defined purity and recommended solvent/formulation guidance for both in vitro and in vivo studies, and is intended for research use only.
  • Potent nicotinamide phosphoribosyltransferase (NAMPT) inhibitor with sub-nanomolar cellular activity.
  • Demonstrated cellular IC50s of 0.7 nM (A2780) and 3.9 nM (COR-L23).
  • Inhibits CYP3A4, enabling metabolic interaction studies.
  • High purity (98.02%) suitable for research applications.
  • Soluble in DMSO and compatible with recommended in vivo formulations.
  • Stable as a powder at -20°C; follow solvent storage guidelines for solutions.