Learn More
AdipoGen Nampt ms rat Dual ELISA Kit
SDP
Supplier:  AdipoGen AG45A0007YEKKI01
Encompass_Preferred
Colorimetric Sandwich ELISA Assay. Detects mouse and rat Nampt. Does not cross-react with human Nampt, mouse adiponectin, mouse resistin, mouse vaspin or mouse RBP4. Colorimetric assay. Sample Types Serum. Range 0.5 to 32ng/ml. Sensitivity 50pg/ml. Nicotinamide phosphoribosyltransferase Nampt pre-B cell colony-enhancing factor PBEF Visfatin is an 52kDA adipokine secreted by adipose tissue and involved in the biosynthesis of nicotinamide adenine dinucleotide NAD. Two forms of Nampt exist, an intracellular form iNampt and an extracellular form eNampt. While the function of iNampt as an essential and rate-limiting NAD biosynthetic enzyme is well established, the physiological role of eNampt is still a matter of debate. It has been shown that Nampt expression is increased in different diseases, like diabetes, obesity, fetal growth retardation, sepsis, inflammatory bowel disease, Crohn’s disease and rheumatoid arthritis.
Catalog No. 50-256-2744