Learn More
AdipoGen Nampt Visfatin ms pAb
SDP
Supplier:  AdipoGen AG25A0028C100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse Nampt. Weakly cross-reacts with human Nampt. Detects a band of ~54kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Nicotinamide phosphoribosyltransferase Nampt pre-B cell colony-enhancing factor PBEF Visfatin is an adipokine that is localized to the bloodstream and has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.
Catalog No. 50-256-3284