Learn More
AdipoGen Nampt Visfatin ms rec
SDP
Supplier:  AdipoGen AG40A0056Y3010
Encompass_Preferred
Protein. Mouse Nampt visfatin/PBEF aa 1-491 is fused at the N-terminus to a FLAG -tag. Source CHO cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >90% SDS-PAGE. Nicotinamide phosphoribosyltransferase Nampt pre-B cell colony-enhancing factor PBEF Visfatin is an adipokine that is localized to the bloodstream and has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.
Catalog No. 50-256-3034