Learn More
Apexbio Technology LLC NAN-190 hydrobromide 115338-32-4 200mg
SDP
Catalog No. 5000091775
Encompass_Preferred
NAN-190 hydrobromide (CAS 115338-32-4) is a selective antagonist of the serotonin 5-HT1A receptor functioning by inhibiting receptor-mediated signal transduction pathways In receptor binding assays NAN-190 hydrobromide demonstrates high affinity for 5-HT1A receptors and modulates neurotransmitter release This compound is frequently employed in neurobiology and psychiatric disorder studies to investigate 5-HT1A-related mechanisms serving as a valuable tool for elucidating serotonergic signaling and its implications in neural function and pathology
Catalog No. 50-000-91775 Supplier Apexbio Technology LLC Supplier No. B6349200
NAN-190 hydrobromide (CAS 115338-32-4) is a selective antagonist of the serotonin 5-HT1A receptor functioning by inhibiting receptor-mediated signal transduction pathways In receptor binding assays NAN-190 hydrobromide demonstrates high affinity for 5-HT1A receptors and modulates neurotransmitter release This compound is frequently employed in neurobiology and psychiatric disorder studies to investigate 5-HT1A-related mechanisms serving as a valuable tool for elucidating serotonergic signaling and its implications in neural function and pathology