Learn More
Medchemexpress LLC NAN-190 hydrobromide | 115338-32-4 | 99.1% | 474.39 | 10 MG
SDP
Catalog No. 5000365258
Encompass_Preferred
NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist, specifically a selective antagonist of 5-HT1A receptors. It is primarily used for research purposes and has been shown to reverse the catalepsy-improving effect of fluoxetine in 6-OHDA lesioned rats.
  • Serotonin receptor 5-HT antagonist
  • Selective antagonist of 5-HT1A receptor
  • Reverses catalepsy-improving effect of fluoxetine
  • For research use only
Catalog No. 50-003-65258 Supplier Medchemexpress LLC Supplier No. 10MGHY19818A
NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist, specifically a selective antagonist of 5-HT1A receptors. It is primarily used for research purposes and has been shown to reverse the catalepsy-improving effect of fluoxetine in 6-OHDA lesioned rats.
  • Serotonin receptor 5-HT antagonist
  • Selective antagonist of 5-HT1A receptor
  • Reverses catalepsy-improving effect of fluoxetine
  • For research use only