Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NAN-190 hydrobromide | 115338-32-4 | 99.1% | 474.39 | 100 MG
SDP

Catalog No. 5000402013
Encompass_Preferred

NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist and a selective antagonist of 5-HT1A. It is intended for research use only.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month

Catalog No. 50-004-02013 Supplier Medchemexpress LLC Supplier No. HY19818A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist and a selective antagonist of 5-HT1A. It is intended for research use only.

  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.