Learn More
Medchemexpress LLC Nan-190 hydrobromide | 115338-32-4 | 99.1% | 474.39 | 200 MG
SDP
Catalog No. 5000401898
Encompass_Preferred
NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist that selectively targets the 5-HT1A receptor. This compound is intended for research use only. In vivo studies demonstrate its ability to reverse the catalepsy-improving effect of fluoxetine in 6-OHDA lesioned rats.
  • Selective antagonist of 5-HT1A receptor
  • Antagonist of 5-HT1 receptor
  • Suitable for research applications
Catalog No. 50-004-01898 Supplier Medchemexpress LLC Supplier No. HY19818A200MG
NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist that selectively targets the 5-HT1A receptor. This compound is intended for research use only. In vivo studies demonstrate its ability to reverse the catalepsy-improving effect of fluoxetine in 6-OHDA lesioned rats.
  • Selective antagonist of 5-HT1A receptor
  • Antagonist of 5-HT1 receptor
  • Suitable for research applications