Learn More
Apexbio Technology LLC Nanaomycin A 52934-83-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061687
Encompass_Preferred
Nanaomycin A is a selective inhibitor targeting DNA methyltransferase 3B (DNMT3B) with an IC50 of approximately 500 nM It acts by specifically inhibiting DNMT3B-mediated DNA methylation without notably affecting DNMT1 activity In cell-based studies employing cancer cell lines such as HCT116 A549 and HL60 treatment with Nanaomycin A reduced global DNA methylation levels and reactivated transcription of tumor suppressor genes previously silenced via promoter hypermethylation Its use in epigenetics-related research includes quantitative assessment of DNMT3B function investigation of DNA methylation regulation mechanisms and evaluation of gene reactivation consequent to DNMT3B inhibition
Catalog No. 50-000-61687 Supplier Apexbio Technology LLC Supplier No. A81915.1
Nanaomycin A is a selective inhibitor targeting DNA methyltransferase 3B (DNMT3B) with an IC50 of approximately 500 nM It acts by specifically inhibiting DNMT3B-mediated DNA methylation without notably affecting DNMT1 activity In cell-based studies employing cancer cell lines such as HCT116 A549 and HL60 treatment with Nanaomycin A reduced global DNA methylation levels and reactivated transcription of tumor suppressor genes previously silenced via promoter hypermethylation Its use in epigenetics-related research includes quantitative assessment of DNMT3B function investigation of DNA methylation regulation mechanisms and evaluation of gene reactivation consequent to DNMT3B inhibition