Learn More
Medchemexpress LLC HY-100528 5mg Medchemexpress, Nanchangmycin CAS:65101-87-3 Purity:>98%
SDP
Catalog No. 502080592
Encompass_Preferred
Medchemexpress, HY-100528 5mg Nanchangmycin CAS:65101-87-3 Nanchangmycin, a polyether antibiotic produced by Streptomyces nanchangensis NS3226, inhibits gram-positive bacteria. Nanchangmycin is a broad spectrum antiviral active against Zika virus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0592 Supplier Medchemexpress LLC Supplier No. HY1005285MG
Medchemexpress, HY-100528 5mg Nanchangmycin CAS:65101-87-3 Nanchangmycin, a polyether antibiotic produced by Streptomyces nanchangensis NS3226, inhibits gram-positive bacteria. Nanchangmycin is a broad spectrum antiviral active against Zika virus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.