Learn More
Educational Insights Nancy B's Science Club™ Way to Grow™ Hydroponics Kit and Gardening Diary
Shop All Learning Resources, Inc Products

Description
- Sprout seeds, put them in the seed baskets, place seed baskets in the water-filled flasks, and watch your plants climb the trellis as they grow
- This set includes everything kids need to explore the amazing world of hydroponics, including 2 seed baskets, 2 flasks with thermometers, and 2 trellises with rulers.
- And the included, 22-page journal is packed with experiments, fun facts, creative writing prompts, and more
- Stimulate an interest in biology and botany with this Nancy B's Science Club set
- Perfect for science fair projects and STEM learning.
Specifications
Specifications
| Grade | K to 12 |
| Class Size | 2 Students |
| Product Type | Gardening Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.