Learn More
Medchemexpress LLC Nangibotide TFA | 99.36% | 1457.46 | 1 MG
SDP
Catalog No. 5000397322
Encompass_Preferred
Nangibotide TFA (LR12 TFA) is a synthetic peptide and a TREM-1 receptor inhibitor. It inhibits NF-κB and NLRP3 inflammasome activation and reduces the release of pro-inflammatory factors such as IL-1β and IL-8. This peptide also inhibits apoptosis, reducing excessive inflammatory responses and protecting tissues from damage. It can be used in research for myocardial ischemia-reperfusion injury, septic shock, acute lung injury, osteoarthritis, and acute liver failure.
  • Inhibits NF-κB and NLRP3 inflammasome activation
  • Reduces release of pro-inflammatory factors like IL-1β and IL-8
  • Protects tissues (liver, lung) from damage
  • Can be used in research for various inflammatory and injury conditions
Catalog No. 50-003-97322 Supplier Medchemexpress LLC Supplier No. HYP3211A1MG
Nangibotide TFA (LR12 TFA) is a synthetic peptide and a TREM-1 receptor inhibitor. It inhibits NF-κB and NLRP3 inflammasome activation and reduces the release of pro-inflammatory factors such as IL-1β and IL-8. This peptide also inhibits apoptosis, reducing excessive inflammatory responses and protecting tissues from damage. It can be used in research for myocardial ischemia-reperfusion injury, septic shock, acute lung injury, osteoarthritis, and acute liver failure.
  • Inhibits NF-κB and NLRP3 inflammasome activation
  • Reduces release of pro-inflammatory factors like IL-1β and IL-8
  • Protects tissues (liver, lung) from damage
  • Can be used in research for various inflammatory and injury conditions