Learn More
Spherotech NANO BLANK POLYSTYRENE KIT
Catalog No. NC0727969
Encompass_Preferred
Nano Polystyrene Size Standard Kit, 1E6/mL, 0.1-0.3 um, 0.4-0.6 um, 0.7-0.9 um, &1.0-1.9 um, 4x5 mL
Catalog No. NC0727969 Supplier Spherotech Supplier No. NPPS4K
Nano Polystyrene Size Standard Kit, 1E6/mL, 0.1-0.3 um, 0.4-0.6 um, 0.7-0.9 um, &1.0-1.9 um, 4x5 mL