Learn More
Proteintech Group Inc Nano-CaptureLigand™ human IgG/rabbit IgG, Fc-specific VHH, biotinylated
SDP
Supplier:  Proteintech Group Inc SHURBGB1100
Encompass
The Nano-CaptureLigand™ human IgG captures and immobilizes the Fc-fragment of human IgG antibodies to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays. It consists of a biotinylated Nanobody that specifically binds to human IgG antibodies with high affinity and in a site-directed manner.
Catalog No. 50-225-8827