Learn More
Analytical Sales & Services Nano Nest Assembly for Photoredox, for use with Glass Plate
Catalog No. 5000804473
Encompass
Nano Nest Assembly for Photoredox Applications\n\nClosed Bottom, for use with Glass Plate (38499)\n\nIncludes: Top Cover, Bottom Plate, Border Gasket (for glass plate), Aluminum Spacers, Silicone Mats, PFA Film and Screws
Catalog No. 50-008-04473 Supplier Analytical Sales & Services Supplier No. 1626200
Nano Nest Assembly for Photoredox Applications\n\nClosed Bottom, for use with Glass Plate (38499)\n\nIncludes: Top Cover, Bottom Plate, Border Gasket (for glass plate), Aluminum Spacers, Silicone Mats, PFA Film and Screws