Learn More
Analytical Sales & Services Nano Nest Assembly for Photoredox, for use with Plastic Collection Plates
Catalog No. 5000804247
Encompass
Nano Nest Assembly for Photoredox Applications\n\nClosed Bottom, for use with Plastic Collection Plates\n\nIncludes: Top Cover, Bottom Plate, Border Gasket (for plastic plate), Aluminum Spacers, Silicone Mats, PFA Films, Acrylic Riser and Screws
Catalog No. 50-008-04247 Supplier Analytical Sales & Services Supplier No. 1626400
Nano Nest Assembly for Photoredox Applications\n\nClosed Bottom, for use with Plastic Collection Plates\n\nIncludes: Top Cover, Bottom Plate, Border Gasket (for plastic plate), Aluminum Spacers, Silicone Mats, PFA Films, Acrylic Riser and Screws